2-Bromo-4-(bromomethyl)benzonitrile 95%

Supplier: Ambeed
Danger

A593211-1G A593211-5G
77017-964EA 76 USD
77017-964 77017-966
2-Bromo-4-(bromomethyl)benzonitrile 95%
2-Bromo-4-(bromomethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅Br₂N
MDL Number: MFCD09261043
CAS Number: 89892-38-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR