Ethyl 6-(trifluoromethyl)picolinate 95%

Supplier: Ambeed
Warning

A593081-250MG A593081-5G
77035-074EA 22.28 USD
77035-074 77035-076
Ethyl 6-(trifluoromethyl)picolinate 95%
Ethyl 6-(trifluoromethyl)picolinate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD11036140
CAS Number: 1010422-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR