4,4'-Oxybis(1,4-phenylene)diboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Oxydibenzeneboronic acid, (Oxybis(4,1-phenylene))diboronic acid

A592323-100MG A592323-250MG
77017-952EA 83.82 USD
77017-952 77017-954
4,4'-Oxybis(1,4-phenylene)diboronic acid 95%
4,4'-Oxybis(1,4-phenylene)diboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂B₂O₅
MW: 257.84 g/mol
MDL Number: MFCD00433207
CAS Number: 19014-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR