DL-Lithium lactate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
DL-Lactic acid lithium salt, (±)-2-Hydroxypropanoic acid lithium salt, Lithium 2-hydroxypropanoate, (±)-2-Hydroxypropionic acid lithium salt, (±)-Lactic acid lithium salt
A591966-25G
A591966-5G
76992-020EA
13
USD
76992-020
76992-022
DL-Lithium lactate 98%
DL-Lithium lactate
Precaución: for Research Use Only
Formula:
H₃CCH(OH)COOLi MW: 96.01 Melting Pt: 300 |
MDL Number:
MFCD00065399 CAS Number: 867-55-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...