3-Aminothiophene-2-carboxamide 97%

Supplier: Ambeed
Danger

A591837-1G A591837-250MG A591837-5G
77017-948EA 27.59 USD
77017-948 77017-950 77572-890
3-Aminothiophene-2-carboxamide 97%
3-Aminothiophene-2-carboxamide

Precaución: for Research Use Only

Formula: C₅H₆N₂OS
MW: 142.18 g/mol
MDL Number: MFCD00052593
CAS Number: 147123-47-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR