2-Bromo-4-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A591752-100MG A591752-1G A591752-250MG
76992-010EA 28 USD
76992-010 76992-012 76992-014
2-Bromo-4-hydroxybenzonitrile 98%
2-Bromo-4-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrNO
MDL Number: MFCD15527339
CAS Number: 82380-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR