[1-(3-Thienyl)ethyl]amine 95%

Supplier: Ambeed
Danger

Synonyms: 1-(Thiophen-3-yl)ethanamine

A591537-100MG A591537-1G A591537-250MG A591537-5G
77017-940EA 85.94 USD
77017-940 77017-942 77017-944 77017-946
[1-(3-Thienyl)ethyl]amine 95%
[1-(3-Thienyl)ethyl]amine

Precaución: for Research Use Only

Formula: C₆H₉NS
MW: 127.21 g/mol
MDL Number: MFCD08452688
CAS Number: 118488-08-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR