Dimethyl 6,6'-dimethyl-[2,2'-bipyridine]-4,4'-dicarboxylate 95%

Supplier: Ambeed
Warning

A591138-100MG A591138-1G A591138-250MG
77017-932EA 51 USD
77017-932 77017-934 77017-936
Dimethyl 6,6'-dimethyl-[2,2'-bipyridine]-4,4'-dicarboxylate 95%
Dimethyl 6,6'-dimethyl-[2,2'-bipyridine]-4,4'-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₄
MDL Number: MFCD00233315
CAS Number: 117330-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR