Chloro[tris(2,4-di-tert-butylphenyl)phosphite]gold 98%

Supplier: Ambeed
Warning

A590192-1G A590192-250MG
76992-000EA 125.19 USD
76992-000 76992-002
Chloro[tris(2,4-di-tert-butylphenyl)phosphite]gold 98%
Chloro[tris(2,4-di-tert-butylphenyl)phosphite]gold

Precaución: for Research Use Only

Formula: C₄₂H₆₃AuClO₃P
MW: 879.34 g/mol
MDL Number: MFCD11045473
CAS Number: 915299-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR