N-Methylmorpholine N-oxide monohydrate 98%

Supplier: Ambeed
Danger

Synonyms: 4-Methylmorpholine N-oxide monohydrate, 4-Methylmorpholine 4-oxide hydrate, NMMO

A590040-100G A590040-25G
76991-996EA 44.56 USD
76991-996 76991-998
N-Methylmorpholine N-oxide monohydrate 98%
N-Methylmorpholine N-oxide monohydrate

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂·1H₂O
MW: 135.16 g/mol
MDL Number: MFCD00149388
CAS Number: 70187-32-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR