2,6-Dimethyl-4-fluorobenzaldehyde 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Fluoro-3-formyl-m-xylene, 4-Fluoro-2,6-dimethylbenzaldehyde
A589901-1G
A589901-250MG
76991-994EA
194.15
USD
76991-994
77684-454
2,6-Dimethyl-4-fluorobenzaldehyde 98+%
2,6-Dimethyl-4-fluorobenzaldehyde
Precaución: For research use only.
Formula:
C₉H₉FO MW: 152.166 g/mol |
MDL Number:
MFCD03701054 CAS Number: 925441-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...