3-Bromo-2-ethoxy-5-(trifluoromethyl)pyridine 95+%

Supplier: Ambeed
Warning

A588698-1G A588698-250MG A588698-5G
76991-988EA 79 USD
76991-988 76991-990 76991-992
3-Bromo-2-ethoxy-5-(trifluoromethyl)pyridine 95+%
3-Bromo-2-ethoxy-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrF₃NO
MDL Number: MFCD14698184
CAS Number: 216766-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR