3-Benzyl-1-(tert-butoxycarbonyl)piperidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A588498-100MG A588498-1G A588498-250MG
77035-062EA 55 USD
77035-062 77035-064 77035-066
3-Benzyl-1-(tert-butoxycarbonyl)piperidine-3-carboxylic acid 97%
3-Benzyl-1-(tert-butoxycarbonyl)piperidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₅NO₄
MDL Number: MFCD03840043
CAS Number: 170838-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR