3,4-Dihydroisoquinolin-2(1H)-yl(oxo)acetic acid 95%

Supplier: Ambeed
Danger

Synonyms: 2-(3,4-Dihydroisoquinolin-2(1H)-yl)-2-oxoacetic acid

A588326-10G A588326-1G
77035-058EA 201.58 USD
77035-058 77035-060
3,4-Dihydroisoquinolin-2(1H)-yl(oxo)acetic acid 95%
3,4-Dihydroisoquinolin-2(1H)-yl(oxo)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.22 g/mol
MDL Number: MFCD03750794
CAS Number: 603097-44-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR