3,3,3-Trifluoropropylene carbonate 99%

Supplier: Ambeed
Warning

A588272-1G A588272-5G A588272-25G
77005-322EA 22 USD
77005-322 77005-324 77572-876
3,3,3-Trifluoropropylene carbonate 99%
3,3,3-Trifluoropropylene carbonate

Precaución: for Research Use Only

Formula: C₄H₃F₃O₃
MDL Number: MFCD02183522
CAS Number: 167951-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR