1,1'-Diheptyl-4,4'-bipyridinium dibromide 95%

Supplier: Ambeed
Warning

Synonyms: 1,1'-Diheptyl-[4,4'-bipyridine]-1,1'-diium bromide

A587221-100G A587221-100MG A587221-1G A587221-250MG A587221-25G A587221-25MG A587221-5G
77035-038EA 856 USD
77035-038 77035-040 77035-042 77035-044 77035-046 77035-048 77035-050
1,1'-Diheptyl-4,4'-bipyridinium dibromide 95%
1,1'-Diheptyl-4,4'-bipyridinium dibromide

Precaución: for Research Use Only

Formula: C₂₄H₃₈Br₂N₂
MDL Number: MFCD00011997
CAS Number: 6159-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR