Diethyl (iodomethyl)phosphonate 98%

Supplier: Ambeed
Warning

A587013-100MG A587013-1G A587013-5G A587013-25G
76991-982EA 15.92 USD
76991-982 76991-984 76991-986 77684-452
Diethyl (iodomethyl)phosphonate 98%
Diethyl (iodomethyl)phosphonate

Precaución: For research use only.

Formula: C₅H₁₂IO₃P
MDL Number: MFCD00041386
CAS Number: 10419-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR