Ethyl 1H-1,2,4-triazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A586999-250MG A586999-5G A586999-1G
76991-978EA 12 USD
76991-978 76991-980 77594-780
Ethyl 1H-1,2,4-triazole-5-carboxylate 95%
Ethyl 1H-1,2,4-triazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
MDL Number: MFCD00619109
CAS Number: 64922-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR