1-Chloro-4,5-difluoro-2-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4,5-difluoronitrobenzene

A586887-1G A586887-25G A586887-5G
76991-970EA 15.92 USD
76991-970 76991-972 76991-974
1-Chloro-4,5-difluoro-2-nitrobenzene 98%
1-Chloro-4,5-difluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₂NO₂
MW: 193.5354 g/mol
MDL Number: MFCD04973764
CAS Number: 771-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR