Sodium 2-ethylhexyl sulfate 40% in water, Ambeed.Inc

Supplier: Ambeed
Warning

A586796-1000G A586796-500G A586796-100G
76991-966EA 43 USD
76991-966 76991-968 77684-451
Sodium 2-ethylhexyl sulfate 40% in water, Ambeed.Inc
Sodium 2-ethylhexyl sulfate

Precaución: For research use only.

Formula: C₈H₁₇NaO₄S
MDL Number: MFCD00042047
CAS Number: 126-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR