2-Methoxy-6-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A586575-1G A586575-250MG A586575-5G
76991-960EA 33 USD
76991-960 76991-962 76991-964
2-Methoxy-6-nitrobenzaldehyde 97%
2-Methoxy-6-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD07371505
CAS Number: 19689-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR