3,5-Dibromo-L-tyrosine 98%

Supplier: Ambeed
Warning

Synonyms: H-Tyr(3,5-DiBr)-OH, H-3,5-Dibromo-Tyr-OH

A585264-1G A585264-5G A585264-25G
77005-318EA 22 USD
77005-318 77602-450 77673-574
3,5-Dibromo-L-tyrosine 98%
3,5-Dibromo-L-tyrosine

Precaución: for Research Use Only

Formula: C₉H₉Br₂NO₃
MW: 338.98 g/mol
MDL Number: MFCD00037204
CAS Number: 300-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR