(S)-tert-Butyl 2-(2,2,2-trifluoroacetyl)pyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A585151-100MG A585151-250MG
76991-950EA 75.33 USD
76991-950 76991-952
(S)-tert-Butyl 2-(2,2,2-trifluoroacetyl)pyrrolidine-1-carboxylate 95%
(S)-tert-Butyl 2-(2,2,2-trifluoroacetyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆F₃NO₃
MW: 267.24 g/mol
MDL Number: MFCD22570647
CAS Number: 913979-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR