PyOxim 95%

Supplier: Ambeed
Danger

Synonyms: (E)-(((1-Cyano-2-ethoxy-2-oxoethylidene)amino)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)

A584913-1G A584913-25G
76991-940EA 6.37 USD
76991-940 76991-942
PyOxim 95%
PyOxim

Precaución: for Research Use Only

Formula: C₁₇H₂₉F₆N₅O₃P₂
MW: 527.389 g/mol
MDL Number: MFCD18643381
CAS Number: 153433-21-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR