2,2'-Bipyridine-4-carboxylic acid 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
[2,2'-Bipyridine]-4-carboxylic acid
A583594-100MG
A583594-1G
A583594-250MG
A583594-5G
77017-894EA
58.35
USD
77017-894
77017-896
77017-898
77684-441
2,2'-Bipyridine-4-carboxylic acid 95%
2,2'-Bipyridine-4-carboxylic acid
Precaución: For research use only.
Formula:
C₁₁H₈N₂O₂ MW: 200.2 g/mol |
MDL Number:
MFCD09414714 CAS Number: 1748-89-6 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...