Furan-2,5-dicarbohydrazide 95%

Supplier: Ambeed
Warning

A583508-100MG A583508-1G
77035-022EA 41.38 USD
77035-022 77035-024
Furan-2,5-dicarbohydrazide 95%
Furan-2,5-dicarbohydrazide

Precaución: for Research Use Only

Formula: C₆H₈N₄O₃
MDL Number: MFCD00612475
CAS Number: 26095-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR