(4-(tert-Butyl)thiazol-2-yl)methanol 95%
Supplier: Ambeed
|
Warning
|
A582660-100MG
A582660-1G
A582660-250MG
77017-888EA
67.9
USD
77017-888
77017-890
77017-892
(4-(tert-Butyl)thiazol-2-yl)methanol 95%
(4-(tert-Butyl)thiazol-2-yl)methanol
Precaución: for Research Use Only
Formula:
C₈H₁₃NOS MW: 171.26 g/mol |
MDL Number:
MFCD12827486 CAS Number: 475059-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...