trans-2-Methyl-2-pentenoic acid 99%

Supplier: Ambeed
Danger

Synonyms: (E)-2-Methylpent-2-enoic acid

A582598-10G A582598-25G
77005-306EA 6.37 USD
77005-306 77005-308
trans-2-Methyl-2-pentenoic acid 99%
trans-2-Methyl-2-pentenoic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00002655
CAS Number: 16957-70-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR