4-Methoxybenzyl 2,2,2-trichloroacetimidate 96%

Supplier: Ambeed
Warning

A582324-5G A582324-1G
77017-886EA 113.01 USD
77017-886 77891-157
4-Methoxybenzyl 2,2,2-trichloroacetimidate 96%
4-Methoxybenzyl 2,2,2-trichloroacetimidate

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₃NO₂
MDL Number: MFCD00134547
CAS Number: 89238-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR