Ethyl 5-bromothiazole-2-carboxylate 98%

Supplier: Ambeed
Warning

A582227-100MG A582227-5G
76991-932EA 47 USD
76991-932 77572-860
Ethyl 5-bromothiazole-2-carboxylate 98%
Ethyl 5-bromothiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MDL Number: MFCD12032197
CAS Number: 1202237-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR