Ethyl 1-methylimidazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A582020-1G A582020-250MG A582020-25G A582020-5G
77035-014EA 35.01 USD
77035-014 77035-016 77035-018 77035-020
Ethyl 1-methylimidazole-4-carboxylate 97%
Ethyl 1-methylimidazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.166 g/mol
MDL Number: MFCD11223488
CAS Number: 41507-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR