3-Hydroxy-3-methylpiperidine 97%

Supplier: Ambeed
Danger

Synonyms: 3-Methylpiperidin-3-ol

A581658-100MG A581658-1G A581658-5G A581658-250MG A581658-25G
77017-878EA 27.59 USD
77017-878 77017-880 77017-882 77684-435 77684-436
3-Hydroxy-3-methylpiperidine 97%
3-Hydroxy-3-methylpiperidine

Precaución: For research use only.

Formula: C₆H₁₃NO
MW: 115.174 g/mol
MDL Number: MFCD09864347
CAS Number: 473730-88-0
UN: 3263
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR