2-(Piperidin-4-yl)ethanol hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A581648-100G
A581648-10G
A581648-1G
A581648-25G
A581648-5G
76991-922EA
971.79
USD
76991-922
76991-924
76991-926
76991-928
76991-930
2-(Piperidin-4-yl)ethanol hydrochloride 98%
2-(Piperidin-4-yl)ethanol hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₆ClNO MW: 165.66 g/mol |
MDL Number:
MFCD14583118 CAS Number: 90747-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...