4'-Bromotri(4-biphenylyl)amine 99%

Supplier: Ambeed
Warning

Synonyms: N,N-Di(4-biphenylyl)-N-(4'-bromo-4-biphenylyl)amine, N,N-Di([1,1'-biphenyl]-4-yl)-4'-bromo-[1,1'-biphenyl]-4-amine

A581437-1G A581437-250MG A581437-5G
77017-872EA 7 USD
77017-872 77017-874 77017-876
4'-Bromotri(4-biphenylyl)amine 99%
4'-Bromotri(4-biphenylyl)amine

Precaución: for Research Use Only

Formula: C₃₆H₂₆BrN
MW: 552.52 g/mol
MDL Number: MFCD22571699
CAS Number: 728039-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR