N-Methyl 4-boronobenzenesulfonamide pinacol ester 98%

Supplier: Ambeed
Danger

Synonyms: N-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide

A581318-1G A581318-250MG A581318-5G
76991-916EA 117.76 USD
76991-916 76991-918 76991-920
N-Methyl 4-boronobenzenesulfonamide pinacol ester 98%
N-Methyl 4-boronobenzenesulphonamide pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₄S
MW: 297.1800 g/mol
MDL Number: MFCD06657895
CAS Number: 1073353-47-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR