2-(3-Methoxyphenoxy)ethanamine 97+%

Supplier: Ambeed
Warning

A581174-1G A581174-250MG A581174-5G
77005-300EA 79 USD
77005-300 77005-302 77005-304
2-(3-Methoxyphenoxy)ethanamine 97+%
2-(3-Methoxyphenoxy)ethanamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MDL Number: MFCD02598966
CAS Number: 6487-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR