3-Fluoro-6-methoxyquinoline 95%

Supplier: Ambeed
Warning

A580164-1G A580164-250MG A580164-5G
76991-904EA 196.27 USD
76991-904 76991-906 76991-908
3-Fluoro-6-methoxyquinoline 95%
3-Fluoro-6-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈FNO
MDL Number: MFCD12400834
CAS Number: 426842-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR