1-(4-Bromophenyl)propan-1-ol 95+%

Supplier: Ambeed
Warning

A579752-1G A579752-250MG A579752-5G
77017-862EA 78 USD
77017-862 77017-864 77891-154
1-(4-Bromophenyl)propan-1-ol 95+%
1-(4-Bromophenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MDL Number: MFCD00021911
CAS Number: 4489-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR