N-(tert-Butoxycarbonyl)-D-proline succinimidyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Pro-OSu, N-Boc-D-proline Succinimidyl Ester

A579636-1G A579636-250MG A579636-25G
77035-008EA 36.08 USD
77035-008 77673-556 77673-558
N-(tert-Butoxycarbonyl)-D-proline succinimidyl ester 97%
N-(tert-Butoxycarbonyl)-D-proline succinimidyl ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₆
MW: 312.32 g/mol
MDL Number: MFCD00069687
CAS Number: 102185-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR