2-Cyclopropyl-6-methylphenol 97%

Supplier: Ambeed
Warning

A579339-1G A579339-250MG
77035-004EA 573 USD
77035-004 77035-006
2-Cyclopropyl-6-methylphenol 97%
2-Cyclopropyl-6-methylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MDL Number: MFCD18072780
CAS Number: 499236-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR