5-Chloro-2-cyclohexyl-1H-benzo[d]imidazole 95+%

Supplier: Ambeed
Warning

A579216-100MG A579216-1G A579216-250MG
77017-848EA 14 USD
77017-848 77017-850 77017-852
5-Chloro-2-cyclohexyl-1H-benzo[d]imidazole 95+%
5-Chloro-2-cyclohexyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₃H₁₅ClN₂
MDL Number: MFCD01929356
CAS Number: 124035-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR