2-Bromo-1,3-dichloro-5-methoxybenzene 98%

Supplier: Ambeed
Warning

A579206-1G A579206-250MG
76991-900EA 160.2 USD
76991-900 76991-902
2-Bromo-1,3-dichloro-5-methoxybenzene 98%
2-Bromo-1,3-dichloro-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
MW: 255.924 g/mol
MDL Number: MFCD26743677
CAS Number: 174913-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR