2-Bromo-1-(difluoromethoxy)-3-fluorobenzene 95%
Supplier: Ambeed
|
Warning
|
A579043-100MG
A579043-1G
A579043-250MG
77017-838EA
14
USD
77017-838
77017-840
77017-842
2-Bromo-1-(difluoromethoxy)-3-fluorobenzene 95%
2-Bromo-1-(difluoromethoxy)-3-fluorobenzene
Precaución: for Research Use Only
Formula:
C₇H₄BrF₃O |
MDL Number:
MFCD18390895 CAS Number: 1239492-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...