4,7-Dibromo-2-octyl-2H-benzo[d][1,2,3]triazole 95%

Supplier: Ambeed
Warning

A578847-100MG A578847-250MG
77017-834EA 46.68 USD
77017-834 77017-836
4,7-Dibromo-2-octyl-2H-benzo[d][1,2,3]triazole 95%
4,7-Dibromo-2-octyl-2H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₁₄H₁₉Br₂N₃
MW: 389.13 g/mol
MDL Number: MFCD19440949
CAS Number: 960509-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR