1-Benzothien-7-ylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: Benzo[b]thiophen-7-ylboronic acid

A578521-100MG A578521-1G A578521-250MG
77034-998EA 29.71 USD
77034-998 77035-000 77035-002
1-Benzothien-7-ylboronic acid 97%
1-Benzothien-7-ylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BO₂S
MW: 178.02 g/mol
MDL Number: MFCD06740320
CAS Number: 628692-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR