4-Amino-N-(2-chlorophenyl)benzamide 95+%

Supplier: Ambeed
Warning

A578423-5G A578423-1G A578423-250MG
77034-996EA 106 USD
77034-996 77673-552 77882-381
4-Amino-N-(2-chlorophenyl)benzamide 95+%
4-Amino-N-(2-chlorophenyl)benzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClN₂O
MDL Number: MFCD00628973
CAS Number: 888-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR