1-Bromo-4-(1-bromoethyl)benzene 95%
Supplier: Ambeed
|
Danger
|
A578020-100MG
A578020-1G
77017-832EA
45.62
USD
77017-832
77684-431
1-Bromo-4-(1-bromoethyl)benzene 95%
1-Bromo-4-(1-bromoethyl)benzene
Precaución: For research use only.
Formula:
C₈H₈Br₂ MW: 263.96 g/mol |
MDL Number:
MFCD11103132 CAS Number: 24308-78-9 UN: 1760 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...