Ethyl 2,4-dihydroxy-5,6-dimethylnicotinate 96%
Supplier: Ambeed
|
Warning
|
A577272-100MG
A577272-1G
A577272-250MG
A577272-5G
76991-868EA
72.15
USD
76991-868
76991-870
76991-872
76991-874
Ethyl 2,4-dihydroxy-5,6-dimethylnicotinate 96%
Ethyl 2,4-dihydroxy-5,6-dimethylnicotinate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₄ MW: 211.21 g/mol |
MDL Number:
MFCD17926290 CAS Number: 77629-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...