4,8-Dichloro-1,7-naphthyridine 95%

Supplier: Ambeed
Warning

A577269-100MG A577269-250MG
76991-864EA 131.56 USD
76991-864 76991-866
4,8-Dichloro-1,7-naphthyridine 95%
4,8-Dichloro-1,7-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂
MW: 199.03 g/mol
MDL Number: MFCD18432768
CAS Number: 1279894-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR