Methyl 5-bromo-1H-imidazole-4-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A576869-100MG
A576869-1G
A576869-250MG
A576869-5G
76991-858EA
19
USD
76991-858
76991-860
76991-862
77886-400
Methyl 5-bromo-1H-imidazole-4-carboxylate 95%
Methyl 5-bromo-1H-imidazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₅H₅BrN₂O₂ |
MDL Number:
MFCD21364526 CAS Number: 1093261-46-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...